Merck
Controlled synthesis of spinel ZnFe2O4 decorated ZnO heterostructures as peroxidase mimetics for enhanced colorimetric biosensing.
Chemical communications (Cambridge, England) (2013-07-23)
Minggang Zhao, Jingyun Huang, Yu Zhou, Xinhua Pan, Haiping He, Zhizhen Ye, Xiaoqing Pan
ABSTRAKT
Controlled synthesis of spinel ZnFe2O4 nanoparticle-decorated ZnO nanofiber heterostructures was carried out with regularly varied particle sizes and uniform distribution. The obtained heterostructures possessed enhanced intrinsic peroxidase-like activity, and the activities regularly changed as the sizes of ZnFe2O4 nanoparticles varied. The heterostructures were further applied in sensitive colorimetric biosensing of urine glucose.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Zinc oxide, 99.99% trace metals basis
Sigma-Aldrich
Zinc oxide, dispersion, nanoparticles, <100 nm particle size (TEM), ≤40 nm avg. part. size (APS), 20 wt. % in H2O
Supelco
Zinc oxide, analytical standard
Sigma-Aldrich
Zinc oxide, 99.999% trace metals basis
Sigma-Aldrich
Zinc oxide, nanopowder, <50 nm particle size (BET), >97%
Sigma-Aldrich
Zinc oxide, nanopowder, ≤50 nm particle size
Sigma-Aldrich
Zinc oxide, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Zinc oxide, ReagentPlus®, powder, <5 μm particle size, 99.9%
Sigma-Aldrich
Zinc oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. for dried substance)
Sigma-Aldrich
Iron(II) oxide, −10 mesh, ≥99.6% trace metals basis