Merck
Responses of state Medicaid programs to buprenorphine diversion: doing more harm than good?
JAMA internal medicine (2013-07-24)
Robin E Clark, Jeffrey D Baxter
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Buprenorphine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Buprenorphine hydrochloride solution, analytical standard, for drug analysis
Supelco
Buprenorphine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®