Merck
Interstitial lung disease due to fumes from heat-cutting polymer rope.
Occupational medicine (Oxford, England) (2013-07-25)
P Sharman, R Wood-Baker
ABSTRAKT
Interstitial lung disease (ILD) due to inhalation of fume/smoke from heating or burning of synthetic polymers has not been reported previously. A fish farm worker developed ILD after cutting rope (polypropylene and nylon) for about 2 hours per day over an extended period using an electrically heated 'knife'. This process produced fume/smoke that entered the workers breathing zone. No other likely cause was identified. This case suggests that exposure to airborne contaminants generated by the heating or burning of synthetic polymers has the potential to cause serious lung disease.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Polyethylene, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Supelco
Polyethylene, analytical standard, suitable for gel permeation chromatography (GPC), 2,000
Sigma-Aldrich
Polyethylene, Medium density
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylene, High density, melt index 12 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Sigma-Aldrich
Polyethylene, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Sigma-Aldrich
Polyethylene, low density, melt index 25 g/10 min (190°C/2.16kg)
Polyethylene (LDPE), ERM®, certified reference material