Merck
Candidate gene study of macular response to supplemental lutein and zeaxanthin.
Experimental eye research (2013-07-31)
Ekaterina Yonova-Doing, Pirro G Hysi, Cristina Venturini, Katie M Williams, Abhishek Nag, Stephen Beatty, S H Melissa Liew, Clare E Gilbert, Christopher J Hammond
ABSTRAKT
Supplementation with carotenoids is proposed to protect against age-related macular degeneration. There is, however, considerable variability in retinal macular pigment response, which may be due to underlying genetic variation. The purpose of this study was to determine whether genetic factors, which have been previously associated with cross-sectional macular pigment levels in the retina or serum lutein, also influence response to supplementation. To this end we conducted an association study in 310 subjects from the TwinsUK cohort between variants in 8 candidate genes and serum lutein and retinal macular pigment optical density (MPOD) levels before and after supplementation. Four variants were associated with MPOD response to supplementation (p < 0.05): rs11057841 (SCARB1), rs4926339 (RPE65), rs1929841 (ABCA1) and rs174534 (FADS1). We also confirmed previous associations between rs6564851 near BMCO1 (p < 0.001) and rs11057841 within SCARB1 (p = 0.01) and baseline measures of serum lutein; while the latter was also associated with MPOD response, none of the BMCO1 variants were. Finally, there was evidence for association between variants near RPE65 and ELOVL2 and changes in lutein concentration after supplementation. This study is the first to show association between genetic variants and response to carotenoids supplementation. Our findings suggest an important link between MP response and the biological processes of carotenoids transport and fatty acid metabolism.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Xanthophyll, from marigold