Merck
  • A baseline study on levels of polychlorinated dibenzo-p-dioxins, polychlorinated dibenzofurans, non-ortho and mono-ortho PCBs, non-dioxin-like PCBs and polybrominated diphenyl ethers in Northeast Arctic cod (Gadus morhua) from different parts of the Barents Sea.
A baseline study on levels of polychlorinated dibenzo-p-dioxins, polychlorinated dibenzofurans, non-ortho and mono-ortho PCBs, non-dioxin-like PCBs and polybrominated diphenyl ethers in Northeast Arctic cod (Gadus morhua) from different parts of the Barents Sea.
Marine pollution bulletin (2013-07-31)
Kaare Julshamn, Arne Duinker, Marc Berntssen, Bente M Nilsen, Sylvia Frantzen, Kjell Nedreaas, Amund Maage
ABSTRAKT
This study is one of several baseline studies on commercially important Norwegian wild fish species that will provide information concerning metals and persistent organic pollutants (POPs) and food safety. The cod liver is a traditional food product in Norway and a potential source for POPs in the diet. The concentrations of dioxins and furans (PCDD/Fs), dioxin-like PCBs (DL-PCBs), non-dioxin-like PCBs (NDL-PCBs, PCB6) and polybrominated flame retardants (PBDEs) were determined in the liver of 784 individual Northeast Arctic cod caught at 32 positions in the Barents Sea in the period from 2009-2010. In addition, muscle samples from 30 individual cod were analysed for the same substances. The mean concentration of the sum of PCDD/Fs and DL-PCBs for all samples was 14.2 ng TEQ who-2005/kg ww with a variation between 1.0 and 151 ng TEQ/kg ww. The concentrations of POPs in the fillet samples were very low.