Merck
  • Simultaneous determination of nicotine and cotinine in serum using high-performance liquid chromatography with fluorometric detection and postcolumn UV-photoirradiation system.
Simultaneous determination of nicotine and cotinine in serum using high-performance liquid chromatography with fluorometric detection and postcolumn UV-photoirradiation system.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2013-07-31)
Makoto Yasuda, Tatsuhiro Ota, Atsushi Morikawa, Ken-ichi Mawatari, Tomoko Fukuuchi, Noriko Yamaoka, Kiyoko Kaneko, Kazuya Nakagomi
ABSTRAKT
A simple and rapid method for the simultaneous determination of serum nicotine and cotinine using high-performance liquid chromatography (HPLC)-fluorometric detection with a postcolumn ultraviolet-photoirradiation system was developed. Analytes were extracted from alkalinized human serum via liquid-liquid extraction using chloroform. The organic phase was back-extracted with the acidified aqueous phase, and the analytes were directly injected into an ion-pair reversed-phase HPLC system. 6-Aminoquinoline was used as an internal standard. Nicotine, cotinine, and 6-aminoquinoline were separated within 14min. The extraction efficiency of nicotine and cotinine was greater than 91%. The linear range was 0.30-1000ng for nicotine and 0.06-1000ng for cotinine. In serum samples from smokers, the concentrations of nicotine and cotinine were 8-15ng/mL and 156-372ng/mL, respectively.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
6-Aminoquinoline, 98%
Sigma-Aldrich
(−)-Cotinine, ≥98%
Supelco
(−)-Cotinine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®