Merck
Antioxidant activity of phosphorylated exopolysaccharide produced by Lactococcus lactis subsp. lactis.
Carbohydrate polymers (2013-08-06)
Yuxing Guo, Daodong Pan, Yangying Sun, Lingying Xin, Hua Li, Xiaoqun Zeng
ABSTRAKT
Exopolysaccharide (EPS) of Lactococcus lactis subsp. lactis was isolated and purified from MRS culture broth. Phosphorylated exopolysaccharide (P-EPS) was synthesized by using the purified EPS and sodium hexametaphosphate (SHMP). The in vitro and in vivo antioxidant activity of EPS and P-EPS was analyzed. Both EPS and P-EPS displayed superoxide anion (O(2-•)), hydroxyl radical (OH•) and DPPH scavenging activity. Catalase (CAT), superoxide dismutase (SOD) and glutathione peroxidase (GSH-Px) activity increased in serum and the livers of mice treated with EPS and P-EPS, while malondialdehyde (MDA) levels decreased. P-EPS was shown to prevent the progression of D-galactose-induced oxidative stress in hepatocytes in vivo. P-EPS showed stronger in vitro and in vivo antioxidant activity than EPS.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl