Merck
Mechanisms underlying taurine protection against glutamate-induced neurotoxicity.
The Canadian journal of neurological sciences. Le journal canadien des sciences neurologiques (2013-08-24)
Hai-Bo Ye, Hai-Bo Shi, Shan-Kai Yin
ABSTRAKT
Taurine appears to exert potent protections against glutamate (Glu)-induced injury to neurons, but the underlying molecular mechanisms are not fully understood. The possibly protected targets consist of the plasma membrane and the mitochondrial as well as endoplasmic reticulum (ER) membranes. Protection may be provided through a variety of effects, including the prevention of membrane depolarization, neuronal excitotoxicity and mitochondrial energy failure, increases in intracellular free calcium ([Ca2+]i), activation of calpain, and reduction of Bcl-2 levels. These activities are likely to be linked spatially and temporally in the neuroprotective functions of taurine. In addition, events that occur downstream of Glu stimulation, including altered enzymatic activities, apoptotic pathways, and necrosis triggered by the increased [Ca2+]i, can be inhibited by taurine. This review discusses the possible molecular mechanisms of taurine against Glu-induced neuronal injury, providing a better understanding of the protective processes, which might be helpful in the development of novel interventional strategies.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Taurine, ≥98%, FG
Supelco
Taurine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Taurine, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
Taurine, ≥99%
Sigma-Aldrich
Taurine, BioUltra, ≥99.5% (T)
SAFC
Taurine