Merck
Antidepressant efficacy of ketamine in treatment-resistant major depression: a two-site randomized controlled trial.
The American journal of psychiatry (2013-08-29)
James W Murrough, Dan V Iosifescu, Lee C Chang, Rayan K Al Jurdi, Charles E Green, Andrew M Perez, Syed Iqbal, Sarah Pillemer, Alexandra Foulkes, Asim Shah, Dennis S Charney, Sanjay J Mathew
ABSTRAKT
Ketamine, a glutamate N-methyl-d-aspartate (NMDA) receptor antagonist, has shown rapid antidepressant effects, but small study groups and inadequate control conditions in prior studies have precluded a definitive conclusion. The authors evaluated the rapid antidepressant efficacy of ketamine in a large group of patients with treatment-resistant major depression. This was a two-site, parallel-arm, randomized controlled trial of a single infusion of ketamine compared to an active placebo control condition, the anesthetic midazolam. Patients with treatment-resistant major depression experiencing a major depressive episode were randomly assigned under double-blind conditions to receive a single intravenous infusion of ketamine or midazolam in a 2:1 ratio (N=73). The primary outcome was change in depression severity 24 hours after drug administration, as assessed by the Montgomery-Åsberg Depression Rating Scale (MADRS). The ketamine group had greater improvement in the MADRS score than the midazolam group 24 hours after treatment. After adjustment for baseline scores and site, the MADRS score was lower in the ketamine group than in the midazolam group by 7.95 points (95% confidence interval [CI], 3.20 to 12.71). The likelihood of response at 24 hours was greater with ketamine than with midazolam (odds ratio, 2.18; 95% CI, 1.21 to 4.14), with response rates of 64% and 28%, respectively. Ketamine demonstrated rapid antidepressant effects in an optimized study design, further supporting NMDA receptor modulation as a novel mechanism for accelerated improvement in severe and chronic forms of depression. More information on response durability and safety is required before implementation in clinical practice.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
(±)-Ketamine hydrochloride, solid
Supelco
Ketamine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®