Merck
Expanded porphyrin-like structures based on twinned triphenylenes.
The Journal of organic chemistry (2013-08-31)
Hemant Gopee, Xiangfei Kong, Zhiqun He, Isabelle Chambrier, David L Hughes, Graham J Tizzard, Simon J Coles, Andrew N Cammidge
ABSTRAKT
Triphenylene twins are intriguing structures, and those bridged through their 3,6-positions by dipyrromethene units give a new class of macrocycles that can be viewed as rigid, expanded porphyrin derivatives in which coplanarity is enforced in a formally antiaromatic π system. Somewhat surprisingly, however, macrocyclization leads to significant overall stabilization of the dipyrromethene chromophores.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Triphenylene, BCR®, certified reference material
Sigma-Aldrich
Triphenylene, 98%