- New insights into von Hippel-Lindau function highlighted by investigation of the trichloroethylene-induced p.P81S hotspot mutation.
New insights into von Hippel-Lindau function highlighted by investigation of the trichloroethylene-induced p.P81S hotspot mutation.
Journal of the National Cancer Institute (2013-08-31)
Len Neckers, Christopher J Ricketts, W Marston Linehan
PMID23990664
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Trichloroethylene, anhydrous, contains 40 ppm diisopropylamine as stabilizer, ≥99%