Merck
  • Artificial receptors inspired by crystal structures of complexes formed between acyclic receptors and monosaccharides: design, syntheses, and binding properties.
Artificial receptors inspired by crystal structures of complexes formed between acyclic receptors and monosaccharides: design, syntheses, and binding properties.
The Journal of organic chemistry (2013-09-05)
Jan Lippe, Monika Mazik
ABSTRAKT
The binding motifs found in the crystal structures of complexes formed between artificial receptors and monosaccharides, reported previously by our group, have inspired us to design new macrocyclic and acyclic receptors, which were expected to form strong 1:1 complexes with monosaccharides, in particular with β-glucosides, through participation in the formation of CH-π interactions and hydrogen bonds with the sugar substrate. As first representatives of these compounds we have prepared the macrocycles 8-12 and the acyclic molecules 13-16, incorporating two central triethylbenzene units. The new compounds had been designed to bind monosaccharides via interactions of both central benzene rings with the sugar CH groups. Initial binding studies have confirmed the expected favorable binding capabilities of the macrocyclic compounds and indicated interesting binding properties of the acyclic analogues.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pyrimidine, ≥98.0%
Supelco
Pyridine solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Pyridine hydrochloride, 98%
Sigma-Aldrich
Pyridine hydrochloride, purum, ≥98.0% (AT)
Supelco
Pyridine, analytical standard
Sigma-Aldrich
Pyridine hydrobromide, 98%
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%