Merck
Losartan, an angiotensin II type 1 receptor blocker, protects human islets from glucotoxicity through the phospholipase C pathway.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2013-09-07)
Anne-Marie Madec, Roméo Cassel, Séverine Dubois, Sylvie Ducreux, Guillaume Vial, Marie-Agnès Chauvin, Aurélia Mesnier, Karim Chikh, Domenico Bosco, Jennifer Rieusset, Fabien Van Coppenolle, Charles Thivolet
ABSTRAKT
As shown in a large clinical prospective trial, inhibition of the renin-angiotensin system (RAS) can delay the onset of type 2 diabetes in high-risk individuals. We evaluated the beneficial effects of RAS inhibition on β-cell function under glucotoxic conditions. Human islets from 13 donors were cultured in 5.5 mM (controls) or 16.7 mM glucose [high glucose (HG)] for 4 d with or without losartan (5 μM), a selective AT1R blocker, and/or U73122 (2 μM), a selective PLC inhibitor, during the last 2 d. HG induced RAS activation with overexpression of AT1R (P<0.05) and angiotensinogen (P<0.001) mRNAs. HG increased endoplasmic reticulum (ER) stress markers (P<0.001) such as GRP78, sXBP1, and ATF4 mRNAs and Grp78 protein levels (P<0.01). HG also decreased reticular calcium concentration (P<0.0001) and modified protein expressions of ER calcium pumps with reduction of SERCA2b (P<0.01) and increase of IP3R2 (P<0.05). Losartan prevented these deleterious effects and was associated with improved insulin secretion despite HG exposure. AT1R activation triggers the PLC-IP3-calcium pathway. Losartan prevented the increase of PLC β1 and γ1 protein levels induced by HG (P<0.05). U73122 reproduced all the protective effects of losartan. AT1R blockade protects human islets from the deleterious effects of glucose through inhibition of the PLC-IP3-calcium pathway.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phospholipase C from Bacillus cereus, ≥200 units/mg protein
Losartan potassium, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Phospholipase C from Clostridium perfringens (C. welchii), Type I, lyophilized powder, 10-50 units/mg protein
Sigma-Aldrich
U-73122 hydrate, powder
Supelco
Losartan potassium
Sigma-Aldrich
Phospholipase C from Clostridium perfringens (C. welchii), Type XIV, lyophilized powder, ≥150 units/mg protein