Merck
Repeated intravenous doxapram induces phrenic motor facilitation.
Experimental neurology (2013-09-10)
M S Sandhu, K Z Lee, E J Gonzalez-Rothi, D D Fuller
ABSTRAKT
Doxapram is a respiratory stimulant used to treat hypoventilation. Here we investigated whether doxapram could also trigger respiratory neuroplasticity. Specifically, we hypothesized that intermittent delivery of doxapram at low doses would lead to long-lasting increases (i.e., facilitation) of phrenic motor output in anesthetized, vagotomized, and mechanically-ventilated rats. Doxapram was delivered intravenously in a single bolus (2 or 6mg/kg) or as a series of 3 injections (2mg/kg) at 5min intervals. Control groups received pH-matched saline injections (vehicle) or no treatment (anesthesia time control). Doxapram evoked an immediate increase in phrenic output in all groups, but a persistent increase in burst amplitude only occurred after repeated dosing with 2mg/kg. At 60min following the last injection, phrenic burst amplitude was 168±24% of baseline (%BL) in the group receiving 3 injections (P<0.05 vs. controls), but was 103±8%BL and 112±4%BL in the groups receiving a single dose of 2 or 6mg/kg, respectively. Following bilateral section of the carotid sinus nerves, the acute phrenic response to doxapram (2mg/kg) was reduced by 68% suggesting that at low doses the drug was acting primarily via the carotid chemoreceptors. We conclude that intermittent application of doxapram can trigger phrenic neuroplasticity, and this approach might be of use in the context of respiratory rehabilitation following neurologic injury.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Doxapram hydrochloride, European Pharmacopoeia (EP) Reference Standard