Merck
Methamphetamine-induced occludin endocytosis is mediated by the Arp2/3 complex-regulated actin rearrangement.
The Journal of biological chemistry (2013-10-02)
Minseon Park, Hyun-Jung Kim, Brian Lim, Adam Wylegala, Michal Toborek
ABSTRAKT
Methamphetamine (METH) is a drug of abuse with neurotoxic and neuroinflammatory effects, which include disruption of the blood-brain barrier (BBB) and alterations of tight junction protein expression. This study focused on the actin cytoskeletal rearrangement as a modulator of METH-induced redistribution of tight junction protein occludin in brain endothelial cells. Exposure to METH resulted in a shift of occludin localization from plasma membranes to endosomes. These changes were accompanied by activation of the actin-related protein 2/3 (Arp2/3) complex, which stimulates actin polymerization by promoting actin nucleation. In addition, METH-induced coronin-1b phosphorylation diminishes the inhibitory effect of nonphosphorylated coronin-1b on actin nucleation. Blocking actin nucleation with CK-666, a specific inhibitor of the Arp2/3 complex, protected against METH-induced occludin internalization and increased transendothelial monocyte migration. Importantly, treatment with CK-666 attenuated a decrease in occludin levels in brain microvessels and BBB permeability of METH-injected mice. These findings indicate that actin cytoskeletal dynamics is detrimental to METH-induced BBB dysfunction by increasing internalization of occludin.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
S(+)-Methamphetamine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®