Merck
  • Phase II trial to assess the safety and efficacy of clofarabine in combination with low-dose cytarabine in elderly patients with acute myeloid leukemia.
Phase II trial to assess the safety and efficacy of clofarabine in combination with low-dose cytarabine in elderly patients with acute myeloid leukemia.
Annals of hematology (2013-10-02)
David Martínez-Cuadrón, Pau Montesinos, Albert Oriol, Olga Salamero, Belén Vidriales, Juan Bergua, Pilar Herrera, Susanna Vives, Jaime Sanz, Cecilia Carpio, Rebeca Rodríguez-Veiga, Federico Moscardó, Miguel A Sanz
ABSTRAKT
Previous studies have shown that clofarabine plus low-dose cytarabine (LDAC) could induce roughly 60 % of complete remissions (CR) with acceptable toxicity and induction mortality in elderly acute myeloid leukemia (AML) patients not suitable for intensive chemotherapy. The Programa Español de Tratamientos en Hematología group conducted a trial for patients diagnosed with untreated AML aged 60 years and older, using the combination of clofarabine (20 mg/m(2) × 5 days) plus low-dose cytarabine (20 mg/m(2) × 14 days). The protocol was flexible regarding the use of antifungal and antibacterial prophylaxis, and outpatient induction therapy was allowed. Although the planned recruitment goal was 75 patients, only 11 patients were enrolled (median age, 74 years) after observing high toxicity and unacceptable mortality (46 and 73 % at 4 and 8 weeks, respectively). The response assessment showed three CR (27 %), three resistant diseases (27 %), and five induction deaths (46 %). Induction was administered in an outpatient modality in five patients, while antifungal and antibacterial prophylaxis was not given in seven and five patients, respectively. In our context, induction therapy with the combination of clofarabine (20 mg/m(2)) plus LDAC was associated with high toxicity and unacceptable mortality in elderly AML patients, leading to the early interruption of the trial. Tight patients' clinical monitoring, follow-up, and intensive supportive care seem crucial to achieve at least acceptable clinical outcomes in elderly AML patients receiving clofarabine plus LDAC. This trial is registered at www.clinicaltrials.gov as no. NCT01193400.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)
Sigma-Aldrich
Cytosine β-D-arabinofuranoside hydrochloride, crystalline
Sigma-Aldrich
Clofarabine, ≥98% (HPLC)