Merck
  • Assessment of deoxyribonucleic acid fragmentation index, testicular volume, semen parameters, and hormone profile in gonadotropin-treated men with hypogonadotropic hypogonadism.
Assessment of deoxyribonucleic acid fragmentation index, testicular volume, semen parameters, and hormone profile in gonadotropin-treated men with hypogonadotropic hypogonadism.
Urology (2013-10-08)
Hani Hosseinifar, Marjan Sabbaghian, Mohammad Chehrazi, Tahereh Modarresi, Firouz Jannat Alipour, Mohammad Ali Sadighi Gilani
ABSTRAKT
To study the sperm deoxyribonucleic acid (DNA) fragmentation index (DFI), testicular volume, semen parameters, and hormone profile in human chorionic gonadotropin (hCG)- and human menopausal gonadotrophin (hMG)-treated patients with hypogonadotropic hypogonadism (HH) with and without a successful pregnancy. This is a cross sectional study. The study initially included 81 patients with HH and azoospermia at the Infertility Unit of Royan Institute between 2010 and 2012. Fifty-eight of 81 patients achieved >1 × 10(6) sperm/mL during hCG and hMG therapy. These 58 patients were divided into the following 2 groups: 20 patients with HH who achieved pregnancy in response to hCG/hMG (responders, 16 naturally and 4 by intrauterine insemination) and 38 gonadotropin-treated patients with HH with failed pregnancy (nonresponders, 29 naturally, 5 by intrauterine insemination, 1 by in vitro fertilization, and 3 by intracytoplasmic sperm injection). Sperm DNA fragmentation was visualized by terminal deoxynucleotidyl transferase dUTP nick end labeling (TUNEL) assay. Average of DFI (responders: 13.45 ± 0.64; nonresponders: 21.92 ± 0.86), age, body mass index, testis volume semen parameters, and follicle-stimulating hormone, luteinizing hormone, and testosterone levels in the 2 groups were calculated. Cut-off point for DFI was determined by receiver operating curve analysis (17.5%). It was shown that DFI in responders is significantly lower than DFI in nonresponders (P <.001), and duration of hCG and hMG therapy in responders is significantly higher than those of nonresponders (P <.05). DFI could be predictive of conception (P <.001; odds ratio 0.57; 95% confidence interval 0.417-0.778). It can be concluded that despite low sperm quality, especially sperm concentration in these patients, decreasing sperm DNA damage may result in successful fertilization.