Merck
PKM2 isoform-specific deletion reveals a differential requirement for pyruvate kinase in tumor cells.
Cell (2013-10-15)
William J Israelsen, Talya L Dayton, Shawn M Davidson, Brian P Fiske, Aaron M Hosios, Gary Bellinger, Jie Li, Yimin Yu, Mika Sasaki, James W Horner, Laura N Burga, Jianxin Xie, Michael J Jurczak, Ronald A DePinho, Clary B Clish, Tyler Jacks, Richard G Kibbey, Gerburg M Wulf, Dolores Di Vizio, Gordon B Mills, Lewis C Cantley, Matthew G Vander Heiden
ABSTRAKT
The pyruvate kinase M2 isoform (PKM2) is expressed in cancer and plays a role in regulating anabolic metabolism. To determine whether PKM2 is required for tumor formation or growth, we generated mice with a conditional allele that abolishes PKM2 expression without disrupting PKM1 expression. PKM2 deletion accelerated mammary tumor formation in a Brca1-loss-driven model of breast cancer. PKM2 null tumors displayed heterogeneous PKM1 expression, with PKM1 found in nonproliferating tumor cells and no detectable pyruvate kinase expression in proliferating cells. This suggests that PKM2 is not necessary for tumor cell proliferation and implies that the inactive state of PKM2 is associated with the proliferating cell population within tumors, whereas nonproliferating tumor cells require active pyruvate kinase. Consistent with these findings, variable PKM2 expression and heterozygous PKM2 mutations are found in human tumors. These data suggest that regulation of PKM2 activity supports the different metabolic requirements of proliferating and nonproliferating tumor cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-PKM2 (isoform M1) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody
Sigma-Aldrich
Pyruvate Kinase from rabbit muscle, Type II, ammonium sulfate suspension, 350-600 units/mg protein
Sigma-Aldrich
Pyruvate Kinase from rabbit muscle, Type VII, buffered aqueous glycerol solution, 350-600 units/mg protein
Sigma-Aldrich
Pyruvate Kinase from rabbit muscle, Type III, lyophilized powder, 350-600 units/mg protein