Merck
Biological evaluation of morin and its new oxovanadium(IV) complex as antioxidant and specific anti-cancer agents.
Chemico-biological interactions (2013-10-16)
Luciana G Naso, Luis Lezama, Teófilo Rojo, Susana B Etcheverry, María Valcarcel, Meritxell Roura, Clarisa Salado, Evelina G Ferrer, Patricia A M Williams
ABSTRAKT
It is known that flavonoids possess, among others, antioxidant and antitumoral properties that depend on their molecular structure. The central objective if this study was to investigate the potential antioxidant and antiproliferative properties of the flavonol morin and its new oxovanadium(IV) complex (VOmor) that was synthesized in order to modify the morin chemical structure. Two osteoblast (UMR106 and MC3T3E1), two breast tumor (T47D and SKBR3) and breast epithelial cell lines in culture were used for the antitumoral determinations. Additionally, a comparative study of their antioxidant capacities using different radicals (DPPH, ABTS(+), OH, O2(-), ROO) was performed. Selected mechanisms of action were studied using the breast cancer cell lines. Results obtained show that morin and its complex behaved as good antioxidant agents for some of the radicals and that the complexation improved the behavior with respect to OH and O2(-) radicals being morin more effective as ROO scavenger. A considerable variation in sensitivity was observed in the breast cancer cells but non-specificity was found for the treatment of osteosarcoma. Moreover, the compounds did not affect the normal proliferation of the breast epithelial mammal cells. The mechanistic studies demonstrated that the complex did not generate reactive oxygen species in the cells (confirming the in vitro studies) and did not produce any damage of DNA. The plasmatic membrane was observed to be damaged only in the SKBR3 cell line. In contrast, the perturbation of the mitochondrial membrane potential and the activation of caspase 3/7 for the breast tumor cells revealed an apoptotic cell death process. All these results collectively suggested that VOmor complex could serve as promising pharmacologically active substance against breast cancer treatment.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis