- Summaries for patients. Screening, monitoring, and treatment of early stage chronic kidney disease: recommendations from the American College of Physicians.
Summaries for patients. Screening, monitoring, and treatment of early stage chronic kidney disease: recommendations from the American College of Physicians.
Annals of internal medicine (2013-10-23)
PMID24146038
MATERIAŁY
Numer produktu
Marka
Opis produktu
Gemfibrozil for system suitability, European Pharmacopoeia (EP) Reference Standard
Gemfibrozil, European Pharmacopoeia (EP) Reference Standard