Merck
No superior performance of hydroxyapatite-coated acetabular cups over porous-coated cups.
Acta orthopaedica (2013-11-01)
Kestutis Valancius, Kjeld Søballe, Poul T Nielsen, Mogens B Laursen
ABSTRAKT
Hydroxyapatite (HA) coating is believed to improve bone-implant ingrowth and long-term survival of prostheses. Recent studies, however, have challenged this view. Furthermore, HA particles may produce third-body wear and initiate aseptic loosening of implants. We report the performance of HA- and porous-coated acetabular cups in a prospective randomized trial. This was an 8-year follow-up study of our previously published prospective randomized study to compare clinical outcomes, survival, periprosthetic bone mineral density, migration, and wear rates of HA- and porous-coated acetabular cups. Dual X-ray absorptiometry (DXA) and Ein Bild Roentgen Analyse (EBRA) measurements were used. 100 patients who underwent unilateral cementless total hip arthroplasty were randomized to either porous-coated cups or HA-coated cups. Patients were examined preoperatively and at 3, 6, and 9 months, and also 1, 3, and 8 years after surgery. 81 patients were available for 8-year follow-up, 40 with porous-coated cups and 41 with HA-coated cups. Age, sex, bone mineral density, and clinical results (Harris hip score) were similar in the 2 groups. The survival, wear, and migration patterns of the cups were also similar in both groups. The results of periprosthetic bone mineral density scans in region of interest 2 was in favor of the porous-coated cups, but there were no differences between the 2 groups in all the remaining regions of interest. HA coating had no statistically significant effect on clinical results, survival, wear, or migration at the 8-year follow-up.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hydroxyapatite, puriss., meets analytical specification of Ph. Eur., BP, FCC, E341, ≥90% (calculated on glowed substance)
Sigma-Aldrich
Hydroxyapatite, purum p.a., ≥90% (as Ca3(PO4)2, KT)