Merck
  • The role of Y84 on domain 1 and Y87 on domain 2 of Paragonimus westermani taurocyamine kinase: Insights on the substrate binding mechanism of a trematode phosphagen kinase.
The role of Y84 on domain 1 and Y87 on domain 2 of Paragonimus westermani taurocyamine kinase: Insights on the substrate binding mechanism of a trematode phosphagen kinase.
Experimental parasitology (2013-11-05)
Blanca R Jarilla, Shinji Tokuhiro, Mitsuru Nagataki, Kouji Uda, Tomohiko Suzuki, Luz P Acosta, Takeshi Agatsuma
ABSTRAKT
The two-domain taurocyamine kinase (TK) from Paragonimus westermani was suggested to have a unique substrate binding mechanism. We performed site-directed mutagenesis on each domain of this TK and compared the kinetic parameters Km(Tc) and Vmax with that of the wild-type to determine putative amino acids involved in substrate recognition and binding. Replacement of Y84 on domain 1 and Y87 on domain 2 with R resulted in the loss of activity for the substrate taurocyamine. Y84E mutant has a dramatic decrease in affinity and activity for taurocyamine while Y87E has completely lost catalytic activity. Substituting H and I on the said positions also resulted in significant changes in activity. Mutation of the residues A59 on the GS region of domain 1 also caused significant decrease in affinity and activity while mutation on the equivalent position on domain 2 resulted in complete loss of activity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
SAFC
L-Tyrosine
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Supelco
L-Tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Supelco
Taurine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Taurine, ≥98%, FG
SAFC
Taurine
Sigma-Aldrich
Taurine, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
Taurine, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Taurine, ≥99%