Merck
Induction regimens for ANCA-Associated Vasculitis.
The New England journal of medicine (2013-11-08)
Ulrich Specks, David Ikle, John H Stone
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Azathioprine, ≥98%
Supelco
Azathioprine, Pharmaceutical Secondary Standard; Certified Reference Material