Merck
  • Clinical results, radiological findings and survival of a proximally hydroxyapatite-coated hip ABG II stem at a minimum of ten years' follow-up: results of a consecutive multicentre study of 1148 hips in 1053 patients.
Clinical results, radiological findings and survival of a proximally hydroxyapatite-coated hip ABG II stem at a minimum of ten years' follow-up: results of a consecutive multicentre study of 1148 hips in 1053 patients.
The bone & joint journal (2013-12-03)
J-A Epinette, G Asencio, J Essig, B Llagonne, C Nourissat
ABSTRAKT
We report a multicentre prospective consecutive study assessing the long-term outcome of the proximally hydroxyapatite (HA)-coated ABG II monobloc femoral component in a series of 1148 hips in 1053 patients with a mean age at surgery of 64.77 years (22 to 80) at a mean follow-up of 10.84 years (10 to 15.25). At latest follow-up, the mean total Harris hip score was 94.7 points (sd; 6.87; 49 to 100), and the mean Merle d'Aubigné-Postel score was 17.6 points (sd 1.12; 7 to 18). The mean total Engh radiological score score was 21.54 (sd 5.77; 3.5 to 27), with 95.81% of 'confirmed ingrowth', according to Engh's classification. With aseptic loosening or pain as endpoints, three AGB II stems (0.26%) failed, giving a 99.7% survival rate (se 0.002; 95% confidence interval (CI) 0.994 to 1) at 14 years' follow-up. The survival of patients ≤ 50 years of age (99.0% (se 0.011; 95% CI 0.969 to 1)) did not differ significantly from those of patients aged > 50 years (99.8% (se 0.002; 95% CI 0.994 to 1)). This study confirmed the excellent long-term results currently achieved with the ABG II proximally HA-coated monobloc stem. Cite this article: Bone Joint J 2013;95-B:1610-16.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hydroxyapatite, puriss., meets analytical specification of Ph. Eur., BP, FCC, E341, ≥90% (calculated on glowed substance)
Sigma-Aldrich
Hydroxyapatite, purum p.a., ≥90% (as Ca3(PO4)2, KT)