Merck
Production of the polysaccharide curdlan by an Agrobacterium strain grown on a plant biomass hydrolysate.
Canadian journal of microbiology (2014-01-08)
Thomas P West, Jessica L Peterson
ABSTRAKT
Production of the commercially available polysaccharide curdlan by Agrobacterium sp. strain ECP-1, isolated as a mutant strain from ATCC 31749, on a medium containing a hydrolysate of the plant prairie cordgrass with selected ammonium phosphate concentrations was investigated for a period of 144 h. Although several ammonium phosphate concentrations supported curdlan production by the strain, the optimal concentration after 120 or 144 h was 3.3 mmol·L⁻¹. Only ammonium phosphate concentrations of 1.1 or 8.7 mmol·L⁻¹ failed to support curdlan production by the strain after 120 or 144 h. Biomass production by strain ECP-1 on the hydrolysate-containing medium after 120 or 144 h was comparable, independent of the ammonium phosphate concentration present. The curdlan yield from the cordgrass hydrolysate indicated that the grass was an effective plant biomass substrate for polysaccharide production.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Curdlan from Alcaligenes faecalis