Merck
A trial of mass isoniazid preventive therapy for tuberculosis control.
The New England journal of medicine (2014-01-24)
Gavin J Churchyard, Katherine L Fielding, James J Lewis, Leonie Coetzee, Elizabeth L Corbett, Peter Godfrey-Faussett, Richard J Hayes, Richard E Chaisson, Alison D Grant
ABSTRAKT
Tuberculosis is epidemic among workers in South African gold mines. We evaluated an intervention to interrupt tuberculosis transmission by means of mass screening that was linked to treatment for active disease or latent infection. In a cluster-randomized study, we designated 15 clusters with 78,744 miners as either intervention clusters (40,981 miners in 8 clusters) or control clusters (37,763 miners in 7 clusters). In the intervention clusters, all miners were offered tuberculosis screening. If active tuberculosis was diagnosed, they were referred for treatment; if not, they were offered 9 months of isoniazid preventive therapy. The primary outcome was the cluster-level incidence of tuberculosis during the 12 months after the intervention ended. Secondary outcomes included tuberculosis prevalence at study completion. In the intervention clusters, 27,126 miners (66.2%) underwent screening. Of these miners, 23,659 (87.2%) started taking isoniazid, and isoniazid was dispensed for 6 months or more to 35 to 79% of miners, depending on the cluster. The intervention did not reduce the incidence of tuberculosis, with rates of 3.02 per 100 person-years in the intervention clusters and 2.95 per 100 person-years in the control clusters (rate ratio in the intervention clusters, 1.00; 95% confidence interval [CI], 0.75 to 1.34; P=0.98; adjusted rate ratio, 0.96; 95% CI, 0.76 to 1.21; P=0.71), or the prevalence of tuberculosis (2.35% vs. 2.14%; adjusted prevalence ratio, 0.98; 95% CI, 0.65 to 1.48; P=0.90). Analysis of the direct effect of isoniazid in 10,909 miners showed a reduced incidence of tuberculosis during treatment (1.10 cases per 100 person-years among miners receiving isoniazid vs. 2.91 cases per 100 person-years among controls; adjusted rate ratio, 0.42; 95% CI, 0.20 to 0.88; P=0.03), but there was a subsequent rapid loss of protection. Mass screening and treatment for latent tuberculosis had no significant effect on tuberculosis control in South African gold mines, despite the successful use of isoniazid in preventing tuberculosis during treatment. (Funded by the Consortium to Respond Effectively to the AIDS TB Epidemic and others; Thibela TB Current Controlled Trials number, ISRCTN63327174.).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Gold, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.05 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <45 μm, 99.99% trace metals basis
Sigma-Aldrich
Gold, rod, diam. 3.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.997% trace metals basis
Sigma-Aldrich
Gold, beads, 1-6 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.3 cm × 0.6 cm, 99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <850 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.025 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold coated glass cover slip, layer thickness 100 Å, 99.999% (Au), L × W × thickness 22 mm × 22 mm × 130-170 μm, square
Sigma-Aldrich
Gold, nanopowder, <100 nm particle size, 99.9% trace metals basis
Sigma-Aldrich
Gold coated glass cover slip, 99.999% (Au), diam. × thickness 15 mm × 130-170 μm, layer thickness 100 Å
Sigma-Aldrich
Gold coated glass slide, layer thickness 100 Å, 99.999% (Au)
Sigma-Aldrich
Gold coated glass slide, layer thickness 1000 Å, 99.999% (Au)
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.99% trace metals basis
Supelco
Isoniazid, analytical standard, ≥99% (TLC)