Merck
Pyrocarbon spacer as a trapezium replacement for arthritis of the trapeziometacarpal joint; a follow-up study of 60 cases.
Acta orthopaedica Belgica (2014-02-26)
Gabor Szalay, Christof Meyer, Tanja Scheufens, Reinhard Schnettler, Ralph Christ, Iris Schleicher
ABSTRAKT
Rhizarthrosis is the most common degenerative joint disease of the hand, affecting about 10% of the population. We report our results with trapezium replacement using a pyrocarbon spacer. Between January 2005 and April 2010, 70 patients underwent trapeziectomy with interposition of a pyrocarbon spacer. Sixty patients were examined at an average follow-up of 23.6 (5-64) months after the operation. Six (8.6%) of the 70 implanted pyrocarbon spacers dislocated. Based on the assessment scale devised by Buck-Gramcko, 19 patients achieved a very good outcome (31.6%), 31 patients (51.6%) had a good outcome, six results were satisfactory (10%) and four patients (6.6%) had a poor result. In this study, trapeziectomy and implantation of a pyrocarbon spacer achieved good or very good results in 83.2% of cases. The high cost of the implant and the observed rate of spacer dislocation should however be considered critically. While the short-term results of this method are encouraging, long-term outcomes will show whether this technique can keep up with the good results of suspension arthroplasty.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Węgiel aktywowany, meets USP testing specifications
Millipore
Activated charcoal, suitable for GC
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Supelco
Activated charcoal, powder
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Sigma-Aldrich
Activated charcoal, DARCO®, −100 mesh particle size, powder