Merck
  • One-pot synthesis of N-methylindoles from N-methylanilines and of benzofurans from phenols using transition-metal carbene X-H insertion reactions.
One-pot synthesis of <I>N</I>-methylindoles from <I>N</I>-methylanilines and of benzofurans from phenols using transition-metal carbene X-H insertion reactions.
Tetrahedron (2009)
Honey MA, et al.