Merck
Gas chromatographic-mass spectrometric quantification of 4-(5-)methylimidazole in roasted coffee after ion-pair extraction.
Journal of chromatography. A (2002-12-05)
S Casal, J O Fernandes, M B P P Oliveira, M A Ferreira
ABSTRAKT
A GC-MS method is described for quantification of 4-(5-)methylimidazole (4MI) in coffee. Although tested, GC-flame ionization detection proved inadequate for this purpose due to the complexity of the coffee matrix. The developed method was based on ion-pair extraction with bis-2-ethylhexylphosphate and derivatization with isobutylchloroformate. Quantification was carried out by the standard addition method using 2-ethylimidazole as internal standard. Reproducibility data from the complete procedure are presented. Mean recoveries were higher than 98%. The method was applied to green and roasted coffee samples from the two most important varieties, arabica and robusta, and to commercial "torrefacto" coffee blends. 4MI was not detected in the green coffee samples analysed and ranged from 0.307 to 1.241 mg/kg in roasted samples.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Ethylimidazole, 98%
Sigma-Aldrich
4(5)-Methylimidazole, 98%