Merck
Identification of the genetic variants of κ-casein in milk by isoelectric focusing electrophoresis.
International Journal of Dairy Technology (2003)
L H Molina, M Casanova, L A Gonzalez, M Pinto, E Carrasco and C Brito