Merck
  • Novel Isatin-Schiff Base Cu (H) and Ni (H) Complexes. X-ray Crystal Structure of Bis [3-(4-hexylphenylimino)-1H-indol-2 (3H)-one]-dichlorocopper (H) Complex.
Novel Isatin-Schiff Base Cu (H) and Ni (H) Complexes. X-ray Crystal Structure of Bis [3-(4-hexylphenylimino)-1H-indol-2 (3H)-one]-dichlorocopper (H) Complex.
Chinese Chemical Letters = Zhongguo Hua Xue Kuai Bao (2006)
Ercag A, et al.