Merck
A transition metal-free Minisci reaction: acylation of isoquinolines, quinolines, and quinoxaline.
The Journal of organic chemistry (2014-04-17)
Yogesh Siddaraju, Manjunath Lamani, Kandikere Ramaiah Prabhu
ABSTRAKT
Transition metal-free acylation of isoquinoline, quinoline, and quinoxaline derivatives has been developed employing a cross dehydrogenative coupling (CDC) reaction with aldehydes using substoichiometric amount of TBAB (tetrabutylammonium bromide, 30 mol %) and K2S2O8 as an oxidant. This intermolecular acylation of electron-deficient heteroarenes provides an easy access and a novel acylation method of heterocyclic compounds. The application of this CDC strategy for acylation strategy has been illustrated in synthesizing isoquinoline-derived natural products.