- Synthesis and Characterization of Two Cyanoxime Ligands, Their Precursors, and Light Insensitive Antimicrobial Silver(I) Cyanoximates.
Synthesis and Characterization of Two Cyanoxime Ligands, Their Precursors, and Light Insensitive Antimicrobial Silver(I) Cyanoximates.
Inorganica chimica acta (2014-04-08)
Courtney N Riddles, Mark Whited, Shalaka R Lotlikar, Korey Still, Marianna Patrauchan, Svitlana Silchenko, Nikolay Gerasimchuk
PMID24707061
ABSTRAKT
High-yield syntheses of N-piperidine-cyanacetamide (