Merck
The chemical constituents of Rhododendron ovatum Planch.
Yao xue xue bao = Acta pharmaceutica Sinica (2005-05-07)
Zi-ming Feng, Ying-hong Wang, Pei-cheng Zhang
ABSTRAKT
To study the chemical constituents of Rhododendron ovatum Planch. The chemical constituents were isolated and purified by silica gel column chromatography and identified on the basis of their physiochemical and spectral data. Seven compounds were isolated and identified. Their structures were established as 3,5,7-trihydroxylchromone 3-O-beta-D-xylopyranoside (I), taraxerol (II), beta-sitosterol (III), betulinic acid (IV), quercetin (V), quercetin-3-O-alpha-L-rhamnopyranoside (VI), and D-glucose (VII). Compound I is a new compound. Compounds II-VII were isolated from this plant for the first time.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
3β-Taraxerol, analytical standard