Merck
[Effect of phenolic alkaloids of Menispermum dauricum on thrombosis and platelet aggregation].
Yao xue xue bao = Acta pharmaceutica Sinica (2006-01-18)
Xiang-Ying Kong, Pei-Li Gong
ABSTRAKT
To observe the effect of phenolic alkaloids of Menispermum dauricum (PAMD) on thrombosis and platelet aggregation, and to explore its mechanism of action. Thrombosis was observed with arteriovenous shunt thrombus model in rat; platelet aggregation was determined by Born's method; ultrastructure of platelet was observed by transmission electron microscope; TXB2 or 6-keto-PGF1alpha levels were assessed by radioimmunoassay; and NO was determined by colorimetric method. PAMD dose-dependently inhibited experimental thrombus formation, platelet aggregation induced by ADP, AA and THR in vivo and ultrastructure changes stimulated by THR; PAMD increased the generation of 6-keto-PGF1alpha in thoracic aortae and NO level in plasma; and had no influence on TXB2 release (P > 0.05). PAMD inhibited thrombosis and platelet aggregation, and its mechanism might be due to the increase of PGI2 and NO level.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Daurisoline, ≥98% (HPLC)