Merck
Synthesis and bronchodilator activity of new quinazolin derivative.
European journal of medicinal chemistry (2006-01-25)
A Zabeer, A Bhagat, O P Gupta, G D Singh, M S Youssouf, K L Dhar, O P Suri, K A Suri, N K Satti, B D Gupta, G N Qazi
ABSTRAKT
Taking lead from a naturally occurring quinazolin vasicine, a number of compounds were developed and evaluated for bronchodilator and anti-allergic activities. One of these compounds was 2,4-diethoxy-6,7,8,9,10,12-hexahydroazepino[2,1-b]quinazolin-12-one, hereinafter named 95-4, exhibited marked bronchodilator activity evaluated on contracted trachea or constricted tracheo-bronchial tree. On intestinal smooth muscle too it showed relaxant effect. Tracheal relaxant effect was not found to be mediated through beta-adrenoceptors. Cumulative dose-response study with acetylcholine and histamine indicated for its non-specific direct effect on smooth muscles. 95-4 was found to be more potent than theophylline and less to that of salbutamol on dose basis. Tested by a number of experimental models, it was found devoid of anti-allergic activity. It was also found to be free from any adverse effect. 95-4 due to its marked bronchial muscle relaxant effect can find use in conditions associated with spasm of bronchial muscles.