Merck
  • Ethyl acetate extract from Angelica Dahuricae Radix inhibits lipopolysaccharide-induced production of nitric oxide, prostaglandin E2 and tumor necrosis factor-alphavia mitogen-activated protein kinases and nuclear factor-kappaB in macrophages.
Ethyl acetate extract from Angelica Dahuricae Radix inhibits lipopolysaccharide-induced production of nitric oxide, prostaglandin E2 and tumor necrosis factor-alphavia mitogen-activated protein kinases and nuclear factor-kappaB in macrophages.
Pharmacological research (2007-01-19)
Ok-Hwa Kang, Go-Hoon Lee, Hyuk Joon Choi, Pil Sang Park, Hee-Sung Chae, Seung-Il Jeong, Youn-Chul Kim, Dong Hwan Sohn, Hyun Park, John Hwa Lee, Dong-Yeul Kwon
ABSTRAKT
Angelica dahurica (Umbelliferae) has been used to treat headache of common cold, supraorbital neuralgia, painful swelling on the body, nasal stuffiness, leukorrhea and arthralgia due to wind-dampness in Korean traditional medicine. It is also claimed to be effective in the treatment of acne, erythema, headache, toothache, sinusitis, colds and flu. The present study focused whether the ethyl acetate extract from Angelica Dahuricae Radix (EAAD) inhibits production of nitric oxide (NO), prostaglandin E(2) (PGE(2)) and tumor necrosis factor (TNF)-alpha, as well as expression of inducible nitric oxide synthase (iNOS), cyclooxygenase-2 (COX-2), nuclear factor-kappaB (NF-kappaB) and mitogen-activated protein kinases (MAPKs) in lipopolysaccharide (LPS)-stimulated macrophages. EAAD inhibited LPS-induced NO, PGE(2) and TNF-alpha production as well as expression of iNOS and COX-2 in RAW 264.7 cells. EAAD inhibited LPS-induced TNF-alpha production in THP-1 cells. Furthermore, EAAD suppressed LPS-induced phosphorylation of p38 MAPK and extracellular-signal regulated kinases 1/2 (ERK1/2), I-kappaBalpha degradation, and NF-kappaB activation in RAW 264.7 cells. These results suggest that EAAD has the inhibitory effects on LPS-induced TNF-alpha, NO and PGE(2) production, and expression of iNOS and COX-2 in macrophage through blockade in the phosphorylation of MAPKs, following I-kappaBalpha degradation and NF-kappaB activation.