Merck
Hypotony maculopathy in a patient with HLA-B27-associated uveitis.
Ocular immunology and inflammation (2008-06-24)
Richard Roe, Bruno Castelo Branco, Emmett T Cunningham
ABSTRAKT
To describe a patient with HLA-B27-associated anterior uveitis who developed hypotony maculopathy. Visual acuity, biomicroscopic slit-lamp examination, and fluorescein angiogram. Following an episode of acute anterior nongranulomatous HLA-B27-associated uveitis, the patient developed decreased vision, choroidal folds, optic disc edema, and a serous exudative detachment characteristic of hypotony maculopathy. The intraocular pressure was noted to be 4 mmHg. Following therapy with topical 1% prednisolone acetate and 5% homatropine, both the intraocular pressure and retinal anatomy normalized. Hypotony maculopathy may be seen in patients with HLA-B27-associated uveitis and should be considered as a cause of vision loss in patients with this condition.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Homatropine hydrobromide, European Pharmacopoeia (EP) Reference Standard