Merck
Diagnosis of bromethalin toxicosis in the dog.
Journal of veterinary diagnostic investigation : official publication of the American Association of Veterinary Laboratory Diagnosticians, Inc (1990-04-01)
D C Dorman, J Simon, K A Harlin, W B Buck
ABSTRAKT
Dogs given a single oral dose of bromethalin at 6.25 mg/kg developed a toxic syndrome characterized by hyperexcitability, tremors, seizures, depression, and death within 15-63 hours after bromethalin administration. Gross lesions included mild cerebral edema (2/5) and mild pulmonary congestion (2/5). Histologic lesions included diffuse white matter spongiosis (5/5), mild microgliosis (3/5), optic nerve vacuolization (3/5), mild thickening of Bowman's capsule (2/5), and occasional splenic megakaryocytes (2/5). Ultramicroscopic examination of midbrain stem revealed occasional swollen axons, intramyelinic vacuolization, and myelin splitting at the intraperiod line. Bromethalin was detected in kidney, liver, fat, and brain tissues, using gas chromatography with electron capture detection. Photodegradation of extracted bromethalin may limit accurate quantification of tissue residues.