Merck
Vitamin A: a multifunctional tool for development.
Seminars in cell & developmental biology (2011-06-23)
Juliana Gutierrez-Mazariegos, Maria Theodosiou, Florent Campo-Paysaa, Michael Schubert
ABSTRAKT
Extensive research carried out over the last 100 years has established that the fat-soluble organic compound vitamin A plays crucial roles in early development, organogenesis, cell proliferation, differentiation and apoptosis as well as in tissue homeostasis. Given its importance during development, the delivery of vitamin A to the embryo is very tightly regulated with perturbations leading to severe malformations. This review discusses the roles of vitamin A during human development and the molecular mechanisms controlling its biological effects, hence bridging the gap between human development and molecular genetic work carried out in animal models. Vitamin A delivery during pregnancy and its developmental teratology in humans are thus discussed alongside work on model organisms, such as chicken or mice, revealing the molecular layout and functions of vitamin A metabolism and signaling. We conclude that, during development, vitamin A-derived signals are very tightly controlled in time and space and that this complex regulation is achieved by elaborate autoregulatory loops and by sophisticated interactions with other signaling cascades.
MATERIAŁY
Numer produktu
Marka
Opis produktu
USP
Acitretin Related Compound B, United States Pharmacopeia (USP) Reference Standard