- Common rodenticide toxicoses in small animals.
Common rodenticide toxicoses in small animals.
The Veterinary clinics of North America. Small animal practice (2012-03-03)
Camille DeClementi, Brandy R Sobczak
PMID22381184
ABSTRAKT
This article focuses on the 3 most commonly used rodenticide types: anticoagulants, bromethalin, and cholecalciferol. It is important to verify the active ingredient in any rodenticide exposure. Many animal owners may use the term “D-con” to refer to any rodenticide regardless of the actual brand name or type of rodenticide. The EPA released their final ruling on rodenticide risk mitigation measures in 2008 and all the products on the market had to be compliant by June 2011, changing to consumer products containing either first-generation anticoagulants or nonanticoagulants including bromethalin and cholecalciferol. These regulations are likely to cause an increase in the number of bromethalin and cholecalciferol cases.