Merck
  • Serotonin 5-HT1A autoreceptor blockade potentiates the ability of the 5-HT reuptake inhibitor citalopram to increase nerve terminal output of 5-HT in vivo: a microdialysis study.
Serotonin 5-HT1A autoreceptor blockade potentiates the ability of the 5-HT reuptake inhibitor citalopram to increase nerve terminal output of 5-HT in vivo: a microdialysis study.
Journal of neurochemistry (1993-02-01)
S Hjorth
ABSTRAKT
The present study addressed the possibility that disinhibition of serotonin (5-HT) autoreceptor-mediated negative feedback might potentiate the elevation of nerve terminal 5-HT output induced by selective 5-HT reuptake blockade. To this end, rats were given citalopram and the 5-HT autoreceptor-blocking agents (S)-UH-301 (5-HT1A) and (-)-penbutolol (5-HT1A/1B), and the effect on extracellular 5-HT in the ventral hippocampus was monitored by means of in vivo microdialysis. Citalopram (5 mg/kg, s.c.) approximately doubled the 5-HT output, a response that was markedly augmented by (S)-UH-301 (3 mg/kg, s.c.) and (-)-penbutolol (8 mg/kg, s.c.) and by combined treatment with (S)-UH-301 (3 mg/kg, s.c.) plus (-)-penbutolol (1 microM; via the dialysis perfusion medium), but not by (-)-penbutolol (1 microM) alone. These findings provide evidence that 5-HT, in particular 5-HT1A, autoreceptor-mediated negative feedback mechanisms are pivotal in determining the nerve terminal 5-HT output level after 5-HT reuptake inhibition. These findings have important implications for the interplay between different processes controlling 5-HT transmission in vivo and might possibly offer a lead toward novel, therapeutically exploitable principles.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Penbutolol sulfate, European Pharmacopoeia (EP) Reference Standard