Merck
Effect of terbium(III) on the binding of aromatic guests with sodium taurocholate aggregates.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2011-07-14)
Tamara C S Pace, Sergio P Souza, Hui Ting Zhang, Cornelia Bohne
ABSTRAKT
The effect of binding Tb(3+) to sodium taurocholate aggregates containing polyaromatic hydrocarbon guests was examined using pyrene and 1-ethylnaphthalene as guests that bind to the primary aggregate, and 1-naphthyl-1-ethanol as a secondary aggregate guest. Time-resolved fluorescence quenching studies were used to study the binding site properties, while laser flash photolysis quenching studies provided information on the dynamics of the guest-aggregate system. Both the primary and secondary aggregate binding sites became more compact in the presence of bound Tb(3+), while only the primary aggregate became more accessible to anionic molecules. The binding dynamics for the guest-primary aggregate system became faster when Tb(3+) was bound to the aggregate. In contrast, for the guest-secondary aggregate the presence of Tb(3+) resulted in a small decrease in the dissociation rate constant. The influence of bound Tb(3+) on the primary and secondary bile salt aggregates is significantly different, which affects how these aggregates can be used as supramolecular host systems to modify guest reactivity.