Merck
  • Stabilization of Analytical Enzymes Using a Novel Polymer-Carbohydrate System and the Production of a Stabilized, Single Reagent for Alcohol Analysis
Stabilization of Analytical Enzymes Using a Novel Polymer-Carbohydrate System and the Production of a Stabilized, Single Reagent for Alcohol Analysis
Analyst (1992)
Timothy D. Gibson, John Higgins, John R. Woodward