Merck
Isolation of fully synthetic promoters for high-level gene expression in Corynebacterium glutamicum.
Biotechnology and bioengineering (2013-05-02)
Sung Sun Yim, Seul Ji An, Misuk Kang, Jinho Lee, Ki Jun Jeong
ABSTRAKT
Corynebacterium glutamicum is an important industrial organism that is widely used in the production of amino acids, nucleotides and vitamins. To extend its product spectrum and improve productivity, C. glutamicum needs to undergo further engineering, including the development of applicable promoter system. Here, we isolated new promoters from the fully synthetic promoter library consisting of 70-bp random sequences in C. glutamicum. Using green fluorescent protein (GFP) as a reporter, highly fluorescent cells were screened from the library by fluorescent activated cell sorting (FACS). Twenty potential promoters of various strengths were isolated and characterized through extensive analysis of DNA sequences and mRNA transcripts. Among 20 promoters, 6 promoters which have different strengths were selected and their activities were successfully demonstrated using two model proteins (antibody fragment and endoxylanase). Finally, the strongest promoter (P(H36)) was employed for the secretory production of endoxylanase in fed-batch cultivation, achieving production levels of 746 mg/L in culture supernatant. This is the first report of synthetic promoters constructed in C. glutamicum, and our screening strategy together with the use of synthetic promoters of various strengths will contribute to the future engineering of C. glutamicum.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Xylanase from Trichoderma viride, lyophilized powder, ≥100 units/mg protein