Merck
Glucose level determination with a multi-enzymatic cascade reaction in a functionalized glass chip.
The Analyst (2013-07-09)
Francesca Costantini, Roald Tiggelaar, Simona Sennato, Francesco Mura, Stefan Schlautmann, Federico Bordi, Han Gardeniers, Cesare Manetti
ABSTRAKT
In this work we show the functionalization of the interior of microfluidic glass chips with poly(2-hydroxyethyl methacrylate) polymer brushes as anchors for co-immobilization of the enzymes glucose-oxidase and horseradish peroxidase. The formation of the brush layer and subsequent immobilization of these enzymes have been characterized on flat surfaces by atomic force microscopy and Fourier transform infrared spectroscopy, and studied inside glass chips by field emission scanning microscopy. Enzyme-functionalized glass chips have been applied for performing a multi-enzymatic cascade reaction for the fast (20 s) determination of glucose in human blood samples and the result is in excellent agreement with values obtained from the conventional hospital laboratory. The limit of detection of this bi-enzymatic method is 60 μM. With the advantages of high selectivity and reproducibility, this functionalization method can be used for improving the efficiency of glucose sensors.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type X-S, lyophilized powder, 100,000-250,000 units/g solid (without added oxygen)
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type VII, lyophilized powder, ≥100,000 units/g solid (without added oxygen)