- Crystallographic and mutational analyses of tannase from Lactobacillus plantarum.
Crystallographic and mutational analyses of tannase from Lactobacillus plantarum.
Proteins (2013-07-10)
Yasuyuki Matoba, Naomi Tanaka, Masafumi Noda, Fumiko Higashikawa, Takanori Kumagai, Masanori Sugiyama
PMID23836494
ABSTRAKT
Tannin acylhydrolase (EC 3.1.1.20) referred commonly as tannase catalyzes the hydrolysis of the galloyl ester bond of tannins to release gallic acid. Although the enzyme is useful for various industries, the tertiary structure is not yet determined. In this study, we determined the crystal structure of tannase produced by Lactobacillus plantarum. The tannase structure belongs to a member of α/β-hydrolase superfamily with an additional "lid" domain. A glycerol molecule derived from cryoprotectant solution was accommodated into the tannase active site. The binding manner of glycerol to tannase seems to be similar to that of the galloyl moiety in the substrate.