Merck
Nuclear PKM2 regulates the Warburg effect.
Cell cycle (Georgetown, Tex.) (2013-09-10)
Weiwei Yang, Zhimin Lu
ABSTRAKT
Pyruvate kinase is a rate-limiting glycolytic enzyme. The PKM1 and PKM2 isoforms result from mutually exclusive alternative splicing of the PKM pre-mRNA. PKM2 rather than PKM1 regulates the Warburg effect and tumorigenesis by poorly understood mechanisms. Emerging evidence has revealed that ERK1/2 phosphorylates PKM2, but not PKM1, leading to PIN1-dependent cis-trans isomerization and conversion of PKM2 from a tetramer to a monomer. Monomeric PKM2 translocates into the nucleus, where it functions as a histone kinase and upregulates the expression of c-Myc and cyclin D1, thereby promoting the Warburg effect and cell cycle progression, respectively. Thus, nuclear PKM2 is essential for tumorigenesis and may serve as a target for treating human cancer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pyruvate Kinase from rabbit muscle, Type II, ammonium sulfate suspension, 350-600 units/mg protein
Sigma-Aldrich
Pyruvate Kinase from rabbit muscle, Type VII, buffered aqueous glycerol solution, 350-600 units/mg protein
Sigma-Aldrich
Pyruvate Kinase from rabbit muscle, Type III, lyophilized powder, 350-600 units/mg protein