Merck
The novel arsenical darinaparsin is transported by cystine importing systems.
Molecular pharmacology (2014-01-17)
Nicolas Garnier, Geneviève G J Redstone, Michael S Dahabieh, Jessica N Nichol, Sonia V del Rincon, Yuxuan Gu, D Scott Bohle, Yan Sun, Douglas S Conklin, Koren K Mann, Wilson H Miller
ABSTRAKT
Darinaparsin (Dar; ZIO-101; S-dimethylarsino-glutathione) is a promising novel organic arsenical currently undergoing clinical studies in various malignancies. Dar consists of dimethylarsenic conjugated to glutathione (GSH). Dar induces more intracellular arsenic accumulation and more cell death than the FDA-approved arsenic trioxide (ATO) in vitro, but exhibits less systemic toxicity. Here, we propose a mechanism for Dar import that might explain these characteristics. Structural analysis of Dar suggests a putative breakdown product: dimethylarsino-cysteine (DMAC). We show that DMAC is very similar to Dar in terms of intracellular accumulation of arsenic, cell cycle arrest, and cell death. We found that inhibition of γ-glutamyl-transpeptidase (γ-GT) protects human acute promyelocytic leukemia cells (NB4) from Dar, but not from DMAC, suggesting a role for γ-GT in the processing of Dar. Overall, our data support a model where Dar, a GSH S-conjugate, is processed at the cell surface by γ-GT, leading to formation of DMAC, which is imported via xCT, xAG, or potentially other cystine/cysteine importing systems. Further, we propose that Dar induces its own import via increased xCT expression. These mechanisms may explain the enhanced toxicity of Dar toward cancer cells compared with ATO.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Arsenic(III) oxide, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Arsenic(III) oxide, 99.995% trace metals basis
Sigma-Aldrich
γ-Glutamyltranspeptidase from porcine kidney, Type IV, 2.0-6.0 units/mg solid
Sigma-Aldrich
γ-Glutamyltranspeptidase from equine kidney, Type VI, 5-12 units/mg solid
Sigma-Aldrich
Arsenic(III) oxide, ReagentPlus®, ≥99.0%
γ-Glutamyl Transferase, ERM®, certified reference material
Sigma-Aldrich
Arsenic(III) oxide, ACS reagent (primary standard)