Merck
Hydrolysis of whey lactose by immobilized β-Galactosidase
Brazilian Archives of Biology and Technology (2008)
Marcela Panaro Mariotti; Hideko Yamanaka; Angela Regina Araujo; Henrique Celso Trevisan